TeacherGeek 40-Piece Wheel Set
Catalog #: 470132-428
Supplier: TEACHERGEEK INC
Specifications
- Description:40-Piece wheel set
- Cat. no.:470132-428
- Supplier No.:28504
- No. of Parts:40
Specifications
About this item
Wheel set to expand your TeacherGeek ideas.
The TeacherGeek Wheel/Hubs are uniquely designed for multiple purposes. When used as wheels for vehicles TeacherGeek Tires may be added for additional traction. Splined holes securely hold dowel axles for a non-slip fit. Combined in pairs they become hubs for turbines with the ability to securely hold 2, 3 or 6 blades that can easily be interchanged.
The TeacherGeek Wheel/Hubs are uniquely designed for multiple purposes. When used as wheels for vehicles TeacherGeek Tires may be added for additional traction. Splined holes securely hold dowel axles for a non-slip fit. Combined in pairs they become hubs for turbines with the ability to securely hold 2, 3 or 6 blades that can easily be interchanged.
Customers Who Bought This Also Bought