Optical Fiber, 400 μm
Catalog #: 470003-014
Supplier: OCEAN OPTICS INC.
Specifications
- Description:400 μm Optical Fiber
- Cat. no.:470003-014
- Length:6''
- Supplier No.:QP400-2VIN
Specifications
About this item
This premium grade optical fiber assembly consistently delivers uniform results with minimal signal variance.
- Terminated with precision SMA 905 connectors, this optical fiber connects easily to the Red Tide Spectrometer, light sources, and sampling accessories
- It can act as both illumination and read fiber
Each assembly is protected with a durable silicone coated steel monocoil jacket with a Dacron braid for superior strain relief. The fiber core diameter is 400 μm.
Customers Who Bought This Also Bought