Three-Oscilloscope Probe Kit
Catalog #: 470201-002
Supplier: CAL TEST ELECTRONICS SE
Specifications
- Description:Three-Oscilloscope Probe Kit
- Cat. no.:470201-002
- Supplier No.:CT3033
Specifications
About this item
Designed for use with general purpose oscilloscopes.
This convenient and cost-effective kit contains three high-performance, passive voltage probes ideal for a wide variety of electronic, electrical, medical, and industrial applications.
Choose between a versatile 1×/10× switchable probe, a 1× fixed probe, or a high voltage 100× fixed attenuation probe. All feature low noise figures, durable construction, slim bodies, and unique snap-locking sprung hooks.
Packaging: Kit contents are packaged in a plastic carrying case for convenient transport and storage.
Customers Who Bought This Also Bought