Respirator Signs, National Marker
Catalog #: 94011-752
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Material:PS Vinyl
- Cat. no.:94011-752
- Legend:Respirator Required (Graphic)
- Dimensions:7.6×12.7 cm (3×5")
- Supplier No.:DGA26AP
- Header:Danger
Specifications
About this item
OSHA signs are the perfect way to alert your employees and guests when a hazard is present in your facility
Keep your facility safe, buy an OSHA regulated sign today and prevent an accident tomorrow.
Customers Who Bought This Also Bought