Septa for Serum Bottles, Ace Glass Incorporated
Catalog #: 89077-548
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Septa for Serum Bottles, For Use with Aluminum Seals
- Cat. no.:89077-548
- Supplier No.:12908-60
Specifications
About this item
Septa, white silicone with structurally bonded PTFE facing
Resists syringe coring. For 20mm mouth O.D., these septa have an O.D. of 19mm, and they are 3mm thick. Use with aluminum crimp seals.
Customers Who Bought This Also Bought