Beakers, Pilot Plant, Ace Glass Incorporated
Catalog #: 89054-040
Supplier: Ace Glass
Supplier #: 6228-05
Product available on GSA Advantage®
Small Business Enterprise
Log in for availability
About this item
This pilot plant beaker is used for large sizes for batch operations and mixing large volumes of measure liquids
- Graduated
- Fabricated from borosilicate glass
Customers Who Bought This Also Bought