Black Dragon Zero Powder-Free Nitrile Exam Gloves, Microflex®
Supplier: Microflex
Certificates
About this item
Black nitrile gloves with textured fingertips offer better tactile sensitivity and grip.
- Ambidextrous
- Powder-free
- Beaded cuffs
Unique black color masks stains and acts as a backdrop for powders, solvents, etc.
Packaging: Packaged 1,000 gloves per case, 100 gloves per dispenser box by weight.
Specifications
- Catalog No:
- CA89230-600
- CA89230-602
- CA89230-604
- CA89230-606
- CA89230-608
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Cuff thickness:
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- Finger thickness:
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- Palm thickness:
- 3.5 mil
- 3.5 mil
- 3.5 mil
- 3.5 mil
- 3.5 mil