Order Entry
ContactUsLinkComponent
GlycoLink IP Kit, Thermo Scientific
GlycoLink IP Kit, Thermo Scientific
Catalog #: CAPI88943
Supplier:  Thermo Scientific
CAS Number:  
GlycoLink IP Kit, Thermo Scientific
Catalog #: CAPI88943
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    GlycoLink IP Kit
  • Size:
    25-rxn kit
  • Supplier No.:
    88943
  • Sufficient For:
    25 IP/Co-IP reactions, each using 20µl antibody-coupled resin

Specifications

About this item

Immunoprecipitate using antibodies immobilized by their Fc-region glycosylation.

The Thermo Scientific GlycoLink IP Kit provides components for effective immunoprecipitation (IP and co-IP) based on small-scale, covalent, affinity-resin immobilization of antibodies and other glycoproteins via oxidized sugar groups.

The GlycoLink Kit uses hydrazide-activated UltraLink Resin to immobilize and perform 25 IP or Co-IP assays with different antibodies or glycoprotein ligands. The method is ideal for polyclonal antibodies, which typically have abundant carbohydrates (glycosylation) on their Fc portions. Monoclonal antibodies that contain adequate carbohydrates are also effective. Antibodies immobilized by this hydrazide-to-carbohydrate method have unobstructed antigen-binding sites and optimal purification capability. The prepared glycoprotein or antibody columns can be regenerated and reused at least five times for immunoprecipitation, co-immunoprecipitation or pull-down affinity-purification without significant loss in binding capacity.

Immunoprecipitation followed by electrophoresis and excision of the protein for mass spectrometry or sequencing. Immunoprecipitation and SDS-PAGE analysis of a protein with a molecular weight similar to the antibody’s heavy or light chain. Identification of binding partners of immobilized glycoprotein. Co-immunoprecipitation for determining protein-protein interactions. The GlycoLink IP Kit method results in covalent attachment of oxidized sugar groups of an IP antibody (or any other glycoprotein with oxidizable carbohydrate groups) to the hydrazide resin. The antibody resin is then incubated with the sample that contains the protein antigen of interest, allowing the antibody-antigen complex to form. After washing to remove non-bound components of the sample, the antigen is recovered by dissociation from the antibody with elution buffer supplied in the kit. The entire procedure is performed in a microcentrifuge spin cup, allowing solutions to be fully separated from the resin with a brief centrifugation. Only the antigen is eluted by the procedure, enabling it to be identified and further analyzed without interference from antibody fragments.

Ordering information: Kit contains resin, buffers and reagents for immobilization; desalting columns; IP lysis/wash and elution buffers for immunoprecipitation; microcentrifuge spin columns and collection tubes; and loading buffer for SDS-PAGE. Kit contains UltraLink Hydrazide Resin, 0.5mL; GlycoLink Coupling Buffer, 50mL; Aniline, 0.2mL; Sodium meta-periodate, 0.5g; Wash Buffer, 60mL; Zeba Spin Desalting Columns,0.5mL, 25 columns; IP Lysis/Wash Buffer, 50mL; PBS Pack (makes 500mL), 1 pack; Elution Buffer, pH 2.8, 50mL; Lane Marker Sample Buffer, Non-reducing, (5X), 5mL; Pierce Spin Columns-Screw Caps, 25 columns; Collection Tubes, 2mL, 100 tubes.
Customers Who Bought This Also Bought