Order Entry
ContactUsLinkComponent
Adapter, Distilling, Ace Glass Incorporated
Adapter, Distilling, Ace Glass Incorporated
Catalog #: 89051-452
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
5085 ADAPTER, Distilling
Adapter, Distilling, Ace Glass Incorporated
Catalog #: 89051-452
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Description:
    Distilling Adapter
  • Cat. no.:
    89051-452
  • Supplier No.:
    5085-10

Specifications

About this item

This adapter is designed for connecting a distilling column with a vertical condenser.

  • Top [ST] outer joint
  • Bottom [ST] inner joints
  • Side arm at 75° angle
  • Accommodates thermometer
  • Fabricated from borosilicate glass

The top outer joint of this adapter is a standard taper 10/30 and is designed for a 76mm immersion thermometer. The side arm is at 75 degrees, and the vertical side arm is 17.8cm from the center tube. Bottom inner joints are standard taper 24/40.

Customers Who Bought This Also Bought