Adapter, Offset, Ace-Threds, Ace Glass Incorporated
Catalog #: 89051-324
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:#7 and #15 Ace-Thred Offset Adapter
- Cat. no.:89051-324
- Supplier No.:5102-05
Specifications
About this item
This adapter features an Ace-Thred at the bottom and two smaller Ace-Threds at the top, with one being offset.
- Bottom Ace-Thred
- Two top Ace-Threds, one offset
- Nylon bushings and o-rings not included
- Fabricated from borosilicate glass
Adapter comes with a #15 Ace-Thred at the bottom and two #7 Ace-Threds at the top, with one being offset. Used with a 89063-384 temperature measurement apparatus for a 89058-884 hydrogenation/gas apparatus. Glass only, NOT supplied with 89051-128 nylon bushings or o-rings.
Customers Who Bought This Also Bought