Adapter, PTFE Stopcock, Ace Glass Incorporated
Catalog #: 89051-416
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- ST Joint:24/40
- Description:1:5 PTFE Stopcock Adapter
- Cat. no.:89051-416
- Supplier No.:5210-40
Specifications
About this item
This adapter features a hose connection and PTFE stopcock plug.
- Reinforced [ST] outer joint
- PTFE stopcock plug
- With hose connection
- Fabricated from borosilicate glass
Adapter has a reinforced standard taper outer joint, a hose connection, and 1:5 solid PTFE stopcock plug. Plug bore is 2mm. For stopcock plug, see 89062-770. Use with 3/8-inch or 5/16-inch I.D. tubing, size C hose connection.
Customers Who Bought This Also Bought