Adapter, Glass Stopcock, Ace Glass Incorporated
Catalog #: 89051-414
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- ST Joint:24/40
- Description:Glass Stopcock Adapter
- Cat. no.:89051-414
- Supplier No.:5210-10
Specifications
About this item
This glass stopcock adapter features a reinforced standard taper outer joint and a hose connection.
- Reinforced [ST] outer joint
- With hose connection
- Fabricated from borosilicate glass
The plug bore is 2mm. For stopcock plug, see 89062-754. Use with 3/8-inch or 5/16-inch I.D. tubing, size C hose connection.
Customers Who Bought This Also Bought