Adapter, Vacuum Jacketed, Ace Glass Incorporated
Catalog #: 89051-342
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Vacuum Jacketed Adapter
- Cat. no.:89051-342
- Supplier No.:5140-10
Specifications
About this item
This vacuum jacketed adapter is used as a distilling head for connecting the top of the column with the side condenser.
- Top [ST] inner joint
- Side and bottom [ST] inner joints
- Accommodates thermometer
- Fabricated from borosilicate glass
The top joint is a standard taper 10/30 inner for a 76mm immersion thermometer. Side and bottom joints are standard taper 24/40 inner. Side arm has drip tip.
Customers Who Bought This Also Bought