Adapter, 75 Degree Side Arm, Borosilicate Glass, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This adapter features a 75° side arm.
- Side and bottom ground [ST] inner joints
- Top reinforced ground [ST] outer joint
- Fabricated from borosilicate glass
Adapter comes with ground standard taper inner joints at the bottom and side, and reinforced ground standard taper outer joint at the top.
Customers Who Bought This Also Bought