Adapter, 105 Degree Side Arm, Borosillicate Glass, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This adapter features a 105° side arm.
- Side reinforced [ST] outer joint
- Bottom [ST] inner joint
- Top Ace-Thred joint
- Fabricated from borosilicate glass
Adapter has a reinforced standard taper outer joint on the side, a standard taper inner joint at the bottom, and an Ace-Thred joint at the top. This adapter is not supplied with bushing and o-rings, which must be ordered separately.
Ordering information: Adapter 89051-250 may be used in conjunction with Bushing with O-Ring 89059-226, or with O-Ring 89061-016. Adapter 89051-252 may be used in conjunction with Bushing with O-Ring 89059-220, or with O-Ring 89060-984.
Customers Who Bought This Also Bought