Adapter, 75 Degree Side Arm, Ace Glass Incorporated
Catalog #: 89051-246
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- ST Joint:24/40
- Description:75 Degree Side Arm Adapter
- Cat. no.:89051-246
- Supplier No.:5045-10
Specifications
About this item
This adapter features a side arm at a 75° angle.
- Top and side reinforced [ST] outer joints
- Bottom [ST] inner joint
- With side arm
- Fabricated from borosilicate glass
This adapter comes with reinforced standard taper outer joints at the top and side, and an standard taper inner joint at the bottom. Side arm is at 75 degree angle.
Customers Who Bought This Also Bought