Adapter, 106 Degree Angle, Ace Glass Incorporated
Catalog #: 89064-802
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:160 Degree Angle Adapter
- Cat. no.:89064-802
- Supplier No.:9056-08
Specifications
About this item
This 160° angled adapter is designed to go from angled flask side joints to a vertical position.
- 160° angle
- Top [ST] outer joint
- Bottom [ST] inner joint
- Fabricated from borosilicate glass
Adapter features a standard taper 14/20 outer joint at top, standard taper 14/20 inner joint at bottom.
Customers Who Bought This Also Bought