Tyvek™ Lockout Tags, NMC
Catalog #: 94016-446
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Material:Tyvek
- Size:17.78Dia. cm (7")
- Cat. no.:94016-446
- Legend:Lockout
- Supplier No.:LOTAG2
Specifications
About this item
Rugged tag withstands moisture, chemicals and frequent handling
Made from lightweight pressure-bonded fibers, this tag is abrasion and tear resistant and features a 5/8" grommet.
Customers Who Bought This Also Bought