Nalgene® Probe Adapter Closure, PP, Thermo Scientific
Catalog #: 16225-250
Supplier: Thermo Fisher Scientific
Specifications
- Cap Size:38-430
- Material:PP
- Supplier No.:2145-0384
Specifications
About this item
Nalgene Probe Adapter Closures allow for insertion of 7- to 14mm diameter probes into Thermo Scientific Nalgene Culture Vessels and Bottles with 38-430 neck finishes
The probe adapter proves a seal between the external environment and the interior of the culture vessels to prevent contamination. The adapter includes a silicone gasket to ensure a leak-proof seal.
Customers Who Bought This Also Bought