Precoiled Pipe Markers 'Caustic', Black/Yellow, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
Certificates
About this item
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007
Customers Who Bought This Also Bought