Precoiled Pipe Markers- 'Drain' W/G, National Marker
Supplier: National Marker
Certificates
About this item
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Customers Who Bought This Also Bought