Glove Box, Closed-Loop Containment, PLAS-LABS™
Supplier: Plas-Labs
Certificates
About this item
This Closed-Loop Containment glove box minimizes drafts and maximizes effectiveness of your balance
Product Benefits: Removable top section, four purging gas valves, white ambidextrous gloves, hospital grade outlet strip, transfer chamber 12" long x 11" ID.
Basic features: Two (2) 8" round HEPA filters, blower, outlet strip, Magnehelic gauges.
This unit features cleanliness, safety, complete containment, portability, and a draft-free atmosphere.The Compact Glove Box is designed specifically for use in hazardous situations where materials to be weighed must be contained.
Certifications: UL Listed.
Ordering information: Glove box comes ready to use, no installation required. Accessory kit includes plug and case.
Specifications
- Catalog No:
- 89218-120
- 89218-122
- 89218-124
- Volume:
- 317 L (11.2 cu. ft.)
- 498 L (17.6 cu. ft.)
- 730 L (25.8 cu. ft.)
- W×D×H ext.:
- 109×61×109 cm (43×24×43")
- 124.5×76.2×109 cm (49×30×43")
- 160×78.7×116.8 cm (63×31×46")
- W×D×H int.:
- 71.1×58.4×73.7 cm (28×23×29")
- 89×73.7×76.2 cm (35×29×30")
- 122×73.7×81.3 cm (48×29×32")
- Electrical:
- 110V
- 110V
- 110V
Specifications
Customers Who Bought This Also Bought