STC1 Series Steam Through Connector, Quick Disconnect Coupling for Bio and Pharma, Colder Products Company
Supplier: Colder Products
Certificates
About this item
Polysulfone connector designed to easily connect stainless steel pharmaceutical process equipment to single use disposable plastic tube sets and media bags
Materials of construction include USP class 6 animal free (BSE/TSE) polysulfone, USP class 6 andimal free (BSE/TSE) silicone orings and seals.
Connect a bioreactor, media bag and steam trap in one easy, sterile connection.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Ordering information: Accessories not included
Customers Who Bought This Also Bought