PRP-X400 Cation Exchange HPLC Columns, Hamilton Company
Supplier: Hamilton
Certificates
About this item
Hamilton Company offers one of the most comprehensive selections of chromatography columns in the industry.
- Detection Levels of Less than 10 ppb when using Post Column, OPA Derivatization Technique
- Applicable to Other Separations, Such as Inositol and Sugar Alcohols
- Three Column Int.Ø. : 2.1 to 4.6 mm
- Two Column Materials: 316 Stainless Steel and PEEK
- Buffer strength 0.0 to 0.5N
Each column is manufactured under rigorous criteria and tested for resolution and retention time accuracy.
The PRP-X400 employs a polymeric cation exchange packing for separation of glyphosate and its metabolite in drinking water. The PRP-X400 column provides a faster separation than the Aminex Glyphosate Column. The PRP-X400 column does not have to be heated to 65 °C, so you don't need to purchase a column heater. PRP-X400 columns cost much less than other glyphosate columns.
Ordering information: Housing Material: Polyetheretherketone (PEEK) and Delrin
Connecting Tubing Size/Material: ¹/₁₆" ext.Ø., 0.25 mm int.Ø./PEEK
Cartridge Materials,
Barrel: PEEK
Frits: Ceramic
Frit Porosity: 2 µm
Cartridge Dimension: 8.0×3.0 mm int.Ø
Cartridge Bed Volume: 56 µl
Particle Size: 12 to 20 µm for Polymeric Packings, 10 µm for Silica Supports
Chemical Compatibility: Recommended for use with virtually all solvents except high concentrations of sodium chloride
Connecting Tubing Size/Material: ¹/₁₆" ext.Ø., 0.25 mm int.Ø./PEEK
Cartridge Materials,
Barrel: PEEK
Frits: Ceramic
Frit Porosity: 2 µm
Cartridge Dimension: 8.0×3.0 mm int.Ø
Cartridge Bed Volume: 56 µl
Particle Size: 12 to 20 µm for Polymeric Packings, 10 µm for Silica Supports
Chemical Compatibility: Recommended for use with virtually all solvents except high concentrations of sodium chloride
Specifications
- Catalog No:
- 89221-092
- 21493-486
- 21493-460
- 21493-093
- 21493-506
- Max. pressure:
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- Temperature limit:
- 5 to 60°C
- 5 to 60°C
- 5 to 60°C
Specifications
Customers Who Bought This Also Bought