Oxygen/Natural Gas Sealing Torch, Labconco®
Catalog #: 26675-851
Supplier: Labconco
Product available on GSA Advantage®
Specifications
- Description:Oxygen/natural gas sealing torch
- Cat. no.:26675-851
- For:Natural gas
- Supplier No.:7578500
Specifications
About this item
Oxygen/Natural Gas Sealing Torch, specifically designed for flame sealing freeze dry ampules
Seals all types of heat-resistant gas. Connects to natural gas, butane or propane and oxygen with 1/4" ID hose connectors.
Customers Who Bought This Also Bought