Blower Transition Adapters, Labconco®
Supplier: Labconco
Certificates
About this item
Epoxy-coated steel blower transition adapters fit all Labconco coated steel blowers
For use with Labconco part nos. 4863500 and 4863501.
Adapter allows you to connect thermoplastic duct to the exhaust side of a blower to create an exhaust stack. A nominal size PVC duct fits inside the opening.
Customers Who Bought This Also Bought