To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Tag
- Antigen name:Green Fluorescence Protein
- Clonality:Monoclonal
- Clone:9F9.F9
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Multiple
- Antigen symbol:GFP
- Conjugation:Texas Red®
- ImmunoFluorescence:Yes
- Size:1 mg
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen synonyms:GFPuv|eGFP|Green Fluorescent Protein
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Anti-Green Fluorescent Protein (GFP) is produced by immunizing GFP containing fusion Protein that corresponds to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria.
- Purification:affinity-purified
- Cat. no.:CA600-309-215
- Supplier No.:600-309-215
Specifications
About this item
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant Proteins. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: 1:500 - 1:2500; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Tag
Antigen: GFP
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: Texas Red®
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Pan