To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin M mu-chain
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgM
- Reactivity:Rat
- Antigen symbol:IgM µ
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:20 mg
- Western blot:Yes
- Epitope:µ-chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:10.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Rat IgM mu heavy chain
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by cross adsorption to remove any unwanted reactivities and extensive dialysis against the buffer stated above
- Cat. no.:CA212-4307
- Supplier No.:212-4307
Specifications
About this item
This product has been assayed against 1.0 µg of Rat IgM in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 2.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Rat