To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Leucine-rich Repeat kinase 2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Antigen symbol:LRRK2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:Center (1200–1300 aa)
- Antigen synonyms:Leucine-rich repeat (LRR) kinase 2|PARK8|Leucine-rich repeat kinase 2
- Cat. no.:60943-030
- Supplier No.:AP7099B
Specifications
About this item
Primary Rabbit Anti-PARK8 (LRRK2) (Center) Reacts with Human, Mouse
Type: Primary
Antigen: LRRK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center (1200–1300 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse