To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Natriuretic Peptide Receptor 1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:NPR1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Epitope:C-Terminal (700–800 aa)
- Antigen synonyms:ANPa|ANPRA|NPRA|GUCY2A|GUC2A
- Cat. no.:80084-930
- Supplier No.:AP8111B
Specifications
About this item
Primary Rabbit Anti-Natriuretic Peptide Receptor A (NPR1/ANPA) (C-term) Reacts with Human
Type: Primary
Antigen: NPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (700–800 aa)
Host: Rabbit
Isotype:
Reactivity: Human