To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:IgG
- Conjugation:Biotin
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:1 mg
- Western blot:Yes
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen synonyms:Fab Anti-Mouse IgG (H&L)
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Mouse IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA810-1602
- Supplier No.:810-1602
Specifications
About this item
Suitable for immunoblotting, ELISA, immunochemistry, immunomicroscopy as well as other antibody based assays using streptavidin or avidin conjugates requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer specificity.
Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse