To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Donkey
- Antigen symbol:IgG
- Conjugation:Biotin
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:2 mg
- Western blot:Yes
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:2.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Donkey IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA616-1602
- Supplier No.:616-1602
Specifications
About this item
Anti-Donkey IgG (H&L) has been assayed against 1.0 µg of Donkey IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Donkey