To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Regulatory Factor X, 5 (Influences HLA Class II Expression)
- Clonality:Polyclonal
- Gene ID:5993
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:RFX5
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µL
- Epitope:near the N-terminus
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:90 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:RFX5 (N-terminal specific) peptide corresponding to a region near the N-terminus of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Purification:delipidation and defibrination
- Cat. no.:CA100-401-193
Specifications
About this item
This product was also tested in a gel supershift assay and found to be reactive against RFX5 complexes using 0.5 to 1.0 µl per assay. This product was also tested in a gel supershift assay and found to be reactive against RFX5 complexes.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: RFX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human