To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent Kinase Inhibitor 1B
- Clonality:Monoclonal
- Clone:5.C3.1
- Gene ID:1027
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:CDKN1B
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:100 µL
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Antigen synonyms:CDKN4|KIP1|MEN1B|P27KIP1 |MEN4 |p27Kip1
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:80 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a full length recombinant human p27 Protein.
- Purification:delipidation and defibrination
- Cat. no.:CA100-401-172
- Supplier No.:100-401-172
Specifications
About this item
This antibody is suitable for use in ELISA, immunoprecipitation, co-immunoprecipitation and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~27 kDa in size corresponding to p27 by western blotting.
Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:1000 - 1:4000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: 5.C3.1
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: