To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Secreted Phosphoprotein 1
- Clonality:Polyclonal
- Gene ID:6696
- Host:Rabbit
- Reactivity:Pig,Human,Dog,Rat,Mouse
- Antigen symbol:SPP1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:200 µL
- Western blot:Yes
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Antigen synonyms:ETA-1|Opnl|Bsp|OP|2AR|BNSP|BSPI|Eta|Ric|Apl-1|Opn|Spp-1|Osteopontin
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:82 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogen:This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide, from the human osteopontin Protein, conjugated to KLH using maleimide.
- Cat. no.:CA100-401-404
- Supplier No.:100-401-404
Specifications
About this item
Anti-Osteopontin is suitable for western blotting, immunochemistry (formalin-fixed paraffin-embedded sections) and ELISA. The antibody exclusively recognizes C-terminal fragments of both thrombin and MMP-cleaved OPN.
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:100 - 1:300; Immunoprecipitation: 1:100;
Type: Primary
Antigen: SPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat, Pig