To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Transcription Factor Y, Alpha
- Clonality:Polyclonal
- Gel shift:Yes
- Gene ID:4800
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NFYA
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µL
- Epitope:near the N-terminus
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Antigen synonyms:CBF-A|CBF-B|NF-YA|HAP2
- Storage buffer:0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:90 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogen:NF-Y (A subunit specific) peptide corresponding to a region near the N-terminus of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Purification:delipidation and defibrination
- Cat. no.:CA100-401-100
Specifications
About this item
Anti-NF-Y (A subunit) Antibody was assayed by immunoblot and found to be reactive against NF-Y (A subunit specific) at a dilution of 1:6,000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).
Recommended Dilutions: ELISA: 1:20000 - 1:100000; GelShift: Use 2.0 µl; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:400;
Type: Primary
Antigen: NFYA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat