To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cullin 1
- Clonality:Polyclonal
- Gene ID:8454
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:CUL1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µL
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Antigen synonyms:Cullin-1
- Amino acid number:766 to 776
- Storage buffer:0.01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 766-776 of Human Cul1 (C-terminus) coupled to KLH.
- Purification:delipidation and defibrination
- Cat. no.:CA100-401-A01
- Supplier No.:100-401-A01
Specifications
About this item
This antibody reacts with human Cul1 by western blot and immunoprecipitation. The antibody immunoprecipitates in vitro translated product and Protein from cell lysates (using HeLa or NIH-3T3).
- Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;
Type: Primary
Antigen: CUL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: