To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:V-myc Avian Myelocytomatosis Viral Oncogene homolog
- Clonality:Polyclonal
- Gene ID:17869
- Host:Rabbit
- Reactivity:Human,Mouse
- Antigen symbol:MYC
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µL
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Antigen synonyms:bHLHe39|Myc2|Nird|AU016757|Niard
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Sequence:KHKLEQLRNSGA
- Storage temperature:–20 °C
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogen:This whole rabbit serum was prepared by repeated immunizations with c-myc peptide corresponding to amino acids KHKLEQLRNSGA which map to the last 12 amino acids of the mouse
- Cat. no.:CA100-401-224
Specifications
About this item
This antibody reacts with human and mouse c-myc Protein, which is a nuclear Protein with a molecular weight of approximately 66 kDa. This antiserum detects c-myc by immunoblotting assays in both overexpressed and endogenous systems.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000;
Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse