To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cullin-associated And Neddylation-dissociated 1
- Clonality:Polyclonal
- Gene ID:55832
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:CAND1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µL
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Antigen synonyms:TIP120|TIP120A
- Amino acid number:11 to 24
- Storage buffer:0.01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 11-24 of Human CAND1/TIP120A (N-terminal) coupled to KLH.
- Purification:delipidation and defibrination
- Cat. no.:CA100-401-A11
- Supplier No.:100-401-A11
Specifications
About this item
This antibody reacts with human, rat and mouse CAND1 by western blot and immunoprecipita-tion. The antibody immunoprecipitates in vitro translated Protein and Protein from cell lysates (using HeLa and NIH-3T3, and others).
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;
Type: Primary
Antigen: CAND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human