To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cyclin T2
- Clonality:Polyclonal
- Gene ID:905
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:CCNT2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Antigen synonyms:CYCT2
- Amino acid number:1 to 584
- Storage buffer:0.01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant Protein corresponding to amino acids 1-584 of human Cyclin T2a Protein.
- Purification:delipidation and defibrination
- Cat. no.:CA100-401-931
- Supplier No.:100-401-931
Specifications
About this item
This antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect bands approximately 74 and 81 kDa in size corresponding to isoforms of Cyclin T2 Protein.
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: CCNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human