To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Alkaline Phosphatase, Intestinal
- Clonality:Polyclonal
- Gene ID:280993
- Host:Rabbit
- Reactivity:Bovine
- Antigen symbol:ALPI
- Conjugation:Unconjugated
- ELISA:Yes
- Size:2 mL
- Western blot:Yes
- Cross adsorption:No
- Format:Antiserum
- Form:Lyophilized
- Storage buffer:0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2,
- Storage temperature:–20 °C
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Ambient
- Immunogen:Alkaline Phosphatase (Calf intestine)
- Purification:delipidation and defibrination
- Cat. no.:CA100-4135
- Supplier No.:100-4135
Specifications
About this item
This product has been assayed against 1.0 µg of Alkaline Phosphatase (Calf intestine) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine