To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Golden syrian hamster
- Antigen symbol:IgG
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:1 mg
- Western blot:Yes
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0
- Storage temperature:+4 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Wet Ice
- Immunogen:Golden Syrian Hamster IgG, whole molecule
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA607-105-130
- Supplier No.:607-105-130
Specifications
About this item
Secondary Goat Anti-IgG (H&L) Reacts with Hamster
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Golden Syrian Hamster