To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Tag
- Antigen name:Green Fluorescence Protein
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Antigen symbol:GFP
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:1 mg
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:GFPuv|eGFP|Green Fluorescent Protein
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.1 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:The immunogen is a Green Fluorescent Protein (GFP) fusion Protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria.
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA600-101-215
- Supplier No.:600-101-215
Specifications
About this item
Anti-GFP is designed to detect GFP and its variants. Goat Anti-GFP is ideal for western blotting, ELISA, immunochemistry and IP.
Recommended Dilutions: ELISA: 1:20000; Immunofluorescence: 1:500; Western Blot: 1:400 - 1:2000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: Yes;
Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Goat
Isotype: IgG
Reactivity: Green Fluorescent Protein