To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:BMI1 Proto-Oncogene, Polycomb Ring Finger
- Clonality:Polyclonal
- Gene ID:648
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:BMI1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:ring finger protein 51|murine leukemia viral (bmi-1) oncogene homolog|flvi-2/bmi-1|polycomb group RING finger protein 4|polycomb group protein Bmi1|RNF51|B lymphoma Mo-MLV insertion region 1 homolog|FLVI2/BMI1|PCGF4|RP11-573G6.1|polycomb complex protein BMI-1
- Amino acid number:252 to 264
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:0.91 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 252-264 of human Bmi1 Protein.
- Purification:affinity-purified
- Cat. no.:CA600-101-392
- Supplier No.:600-101-392
Specifications
About this item
This affinity purified antibody has been tested for use in ELISA and western blot. A 1:200 dilution is reported to be effective for Immunofluorescence against Methanol fixed cells. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:5000 - 1:30,000; Immunofluorescence: 1:200; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: BMI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human