To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Amyloid beta (A4) Precursor Protein
- Clonality:Polyclonal
- Gene ID:351
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:APP
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:0.99mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:This antibody was affinity purified from whole rabbit serum prepared by repeated immunizations with a synthetic peptide corresponding to the amino terminus (aa 1-14) of human beta amyloid conjugated to KLH using maleimide.
- Purification:affinity-purified
- Cat. no.:CA600-401-253
- Supplier No.:600-401-253
Specifications
About this item
Affinity purified anti-beta amyloid detects beta amyloid in ELISA, Immunochemistryand IF. A 40-50 kD band consistent with a higher MW precursor is detected in western blot using whole tissue extracts from mouse brain.
- Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:50-1:200; Western Blot: 1:1000-1:5000; Immunochemistry: 1:50-1:200;
Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse