To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:clusterin
- Clonality:Polyclonal
- Gene ID:1191
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CLU
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:500 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:SGP-2|NA1/NA2|KUB1|SP-40|TRPM-2|SGP2|CLU1|APOJ|CLI|APO-J|TRPM2|AAG4|CLU2
- Storage buffer:0.125 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Wet Ice
- Immunogen:apoLipoProtein Type J was isolated from human plasma by density gradient centrifugation followed by HPLC purification.
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA600-101-198
- Supplier No.:600-101-198
Specifications
About this item
Anti-apoLipoProtein antibodies have been used for indirect trapping ELISA for quantitation of antigen in serum using a standard curve, for immunoprecipitation and for western blotting for highly sensitive qualitative analysis.
Recommended Dilutions: ELISA: 1:5000 - 1:10000; Western Blot: 1:5000 - 1:10000; Immunochemistry: 1:50 - 1:200; Immunoprecipitation: 1:100;
Type: Primary
Antigen: CLU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: