To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ankyrin Repeat Domain 26
- Clonality:Polyclonal
- Gene ID:232339
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:ANKRD26
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:BC008111|mKIAA1074|5730521P14Rik
- Amino acid number:1 to 15
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.48 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 1-15 of mouse Ankrd26 Protein.
- Purification:affinity-purified
- Cat. no.:CA600-401-917
- Supplier No.:600-401-917
Specifications
About this item
This affinity purified antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 81 kDa in size corresponding to Ankrd26 by western blotting.
Recommended Dilutions: ELISA: 1:20000 - 1:80,000; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: ANKRD26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse