To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:AKT Serine/threonine Kinase 1
- Clonality:Polyclonal
- Gene ID:207
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:AKT1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:CWS6|RAC|RAC-ALPHA|PRKBA|PKB|AKT|PKB-ALPHA
- Amino acid number:460 to 480
- Modification:phospho-Ser473
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.02 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide corresponding to the C-terminus aa 460-480 of human, mouse, rat and chicken AKT Proteins conjugated to KLH.
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA600-401-268
- Supplier No.:600-401-268
Specifications
About this item
This antibody is phospho specific for pS473 and is suitable for western blotting, immunochemistry (formalin-fixed paraffin-embedded sections), immunofluorescence and ELISA. By immunoblot a single band of the expected apparent molecular weight is obse
Recommended Dilutions: ELISA: 1:15000 - 1:60,000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:100 - 1:500;
Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat