Specifications
- Description:CytoScan™ SRB Cytotoxicity Assay
- Size:1000 assays
- Cat. no.:CA95029-190
- Supplier No.:786-213
Specifications
About this item
G-Biosciences' CytoScan™ SRB Cytotoxicity Assay is a simple, accurate and reproducible assay based upon the quantitative staining of cellular proteins by sulforhodamine B (SRB).
G-Biosciences' CytoScan™ SRB Cytotoxicity Assay has been used for high-throughput drug screening at the National Cancer Institute. Sulforhodamine B is an anionic aminoxanthene dye that forms an electrostatic complex with the basic amino acid residues of proteins under moderately acid conditions, which provides a sensitive linear response. The color development is rapid and stable and is readily measured at absorbances between 560 and 580nm.
G-Biosciences' CytoScan™ SRB Cytotoxicity Assay is supplied in a kit that contains components sufficient for performing up to 1000 assays.