Specifications
- Description:CytoScan™ WST-1 Cell Proliferation Assay
- Size:500 assays
- Cat. no.:CA95029-188
- Supplier No.:786-212
Specifications
About this item
G-Biosciences' CytoScan™ WST-1 Cell Proliferation Assay is a sensitive and accurate assay for cell proliferation and cytotoxicity
G-Biosciences' CytoScan™ WST-1 Cell Proliferation Assay principle is based upon the reduction of the high sensitivity tetrazolium salt, WST-1, to formazan by cellular dehydrogenases. The generation of the dark yellow colored formazan is measured at 420 to 480nm (optimal at 440nm) and is directly correlated to cell number.
G-Biosciences' CytoScan™ WST-1 Cell Proliferation Assay is supplied in a kit that contains components sufficient for performing up to 500 assays.
The assay is highly convenient as it is performed in a single cell tissue culture well and requires no washing, harvesting or solubilization of cells. Adherent or suspension cells are cultured in a microplate and then incubated with WST-1 and the assay is monitored with a spectrophotometer.